Anti-HTR1B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14575CEA 350 EUR
ABGEAP14575C
Anti-HTR1B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HTR1B (5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR