Anti-SIGLEC8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14564AEA 348 EUR
ABGEAP14564A
Anti-SIGLEC8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SIGLEC8 (sialic acid binding Ig-like lectin 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR