Anti-FDFT1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14561BEA 350 EUR
ABGEAP14561B
Anti-FDFT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FDFT1 (farnesyl-diphosphate farnesyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR