Anti-HSFX1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14550CEA 348 EUR
ABGEAP14550C
Anti-HSFX1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSFX1 (heat shock transcription factor family, X linked 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR