Anti-KCNC3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14547BEA 350 EUR
ABGEAP14547B
Anti-KCNC3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KCNC3 (potassium voltage-gated channel, Shaw-related subfamily, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR