Anti-EPM2A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1453BEA 350 EUR
ABGEAP1453B
Anti-EPM2A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EPM2A (epilepsy, progressive myoclonus type 2A, Lafora disease (laforin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR