Anti-GABRB2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14538BEA 348 EUR
ABGEAP14538B
Anti-GABRB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GABRB2 (gamma-aminobutyric acid (GABA) A receptor, beta 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR