Anti-PTPRN Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14523CEA 360 EUR
ABGEAP14523C
Anti-PTPRN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTPRN (protein tyrosine phosphatase, receptor type, N)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR