Anti-ADAM32 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14513AEA 350 EUR
ABGEAP14513A
Anti-ADAM32 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADAM32 (ADAM metallopeptidase domain 32)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR