Anti-PERP Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14508BEA 350 EUR
ABGEAP14508B
Anti-PERP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PERP (PERP, TP53 apoptosis effector)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR