Anti-GTF2E1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14432AEA 350 EUR
ABGEAP14432A
Anti-GTF2E1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GTF2E1 (general transcription factor IIE, polypeptide 1, alpha 56kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR