Anti-SHMT1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14423AEA 360 EUR
ABGEAP14423A
Anti-SHMT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SHMT1 (serine hydroxymethyltransferase 1 (soluble))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR