Anti-SWAP70 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1441CEA 348 EUR
ABGEAP1441C
Anti-SWAP70 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SWAP70 (SWAP switching B-cell complex 70kDa subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR