Anti-PPEF1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14395CEA 350 EUR
ABGEAP14395C
Anti-PPEF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPEF1 (protein phosphatase, EF-hand calcium binding domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR