Anti-GTF2A1L Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14377BEA 350 EUR
ABGEAP14377B
Anti-GTF2A1L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GTF2A1L (general transcription factor IIA, 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR