Anti-PUF60 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14370AEA 348 EUR
ABGEAP14370A
Anti-PUF60 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PUF60 (poly-U binding splicing factor 60KDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR