Anti-IL9R Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14364CEA 348 EUR
ABGEAP14364C
Anti-IL9R Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL9R (interleukin 9 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR