Anti-RNF40 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14362BEA 350 EUR
ABGEAP14362B
Anti-RNF40 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RNF40 (ring finger protein 40, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR