Anti-SMARCE1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14356BEA 360 EUR
ABGEAP14356B
Anti-SMARCE1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMARCE1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR