Anti-GTF2B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14354AEA 350 EUR
ABGEAP14354A
Anti-GTF2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GTF2B (general transcription factor IIB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR