Anti-ALDH2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1432CEA 360 EUR
ABGEAP1432C
Anti-ALDH2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALDH2 (aldehyde dehydrogenase 2 family (mitochondrial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR