Anti-CDK15 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14286AEA 350 EUR
ABGEAP14286A
Anti-CDK15 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDK15 (cyclin-dependent kinase 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR