Anti-USP43 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14283BEA 360 EUR
ABGEAP14283B
Anti-USP43 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: USP43 (ubiquitin specific peptidase 43)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR