Anti-AKT1S1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14275BEA 350 EUR
ABGEAP14275B
Anti-AKT1S1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKT1S1 (AKT1 substrate 1 (proline-rich))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR