Anti-PDGFRA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14254CEA 348 EUR
ABGEAP14254C
Anti-PDGFRA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PDGFRA (platelet-derived growth factor receptor, alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR