Anti-KCNK10 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14248BEA 350 EUR
ABGEAP14248B
Anti-KCNK10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KCNK10 (potassium channel, subfamily K, member 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR