Anti-TMC8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14243AEA 348 EUR
ABGEAP14243A
Anti-TMC8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMC8 (transmembrane channel-like 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR