Anti-NDUFA13 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14220AEA 350 EUR
ABGEAP14220A
Anti-NDUFA13 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NDUFA13 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR