Anti-CCL15 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14217BEA 350 EUR
ABGEAP14217B
Anti-CCL15 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCL15 (chemokine (C-C motif) ligand 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR