Anti-NR2E3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14205CEA 350 EUR
ABGEAP14205C
Anti-NR2E3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NR2E3 (nuclear receptor subfamily 2, group E, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR