Anti-TCEB3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14198CEA 348 EUR
ABGEAP14198C
Anti-TCEB3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TCEB3 (transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR