Anti-OXSR1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14158AEA 350 EUR
ABGEAP14158A
Anti-OXSR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OXSR1 (oxidative stress responsive 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR