Anti-KSR1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14150BEA 348 EUR
ABGEAP14150B
Anti-KSR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KSR1 (kinase suppressor of ras 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR