Anti-CDKN2B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP14138BEA 350 EUR
ABGEAP14138B
Anti-CDKN2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDKN2B (cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR