Anti-GNL3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1405BEA 350 EUR
ABGEAP1405B
Anti-GNL3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GNL3 (guanine nucleotide binding protein-like 3 (nucleolar))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR