Anti-CDH8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1402AEA 348 EUR
ABGEAP1402A
Anti-CDH8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDH8 (cadherin 8, type 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR