Anti-CDH4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1401AEA 348 EUR
ABGEAP1401A
Anti-CDH4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDH4 (cadherin 4, type 1, R-cadherin (retinal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR