Anti-AP2A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13905BEA 350 EUR
ABGEAP13905B
Anti-AP2A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AP2A1 (adaptor-related protein complex 2, alpha 1 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR