Anti-CHRNA7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13898AEA 348 EUR
ABGEAP13898A
Anti-CHRNA7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHRNA7 (cholinergic receptor, nicotinic, alpha 7 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR