Anti-DEF8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13880BEA 350 EUR
ABGEAP13880B
Anti-DEF8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DEF8 (differentially expressed in FDCP 8 homolog (mouse))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR