Anti-RARA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13849BEA 370 EUR
ABGEAP13849B
Anti-RARA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RARA (retinoic acid receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR