Anti-ABHD3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13830BEA 348 EUR
ABGEAP13830B
Anti-ABHD3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ABHD3 (abhydrolase domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR