Anti-BCL2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13823CEA 360 EUR
ABGEAP13823C
Anti-BCL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BCL2 (B-cell CLL/lymphoma 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR