Anti-POLR3B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13819AEA 350 EUR
ABGEAP13819A
Anti-POLR3B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: POLR3B (polymerase (RNA) III (DNA directed) polypeptide B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR