Anti-MGAT5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13815BEA 350 EUR
ABGEAP13815B
Anti-MGAT5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MGAT5 (mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR