Anti-OR1N1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13812BEA 350 EUR
ABGEAP13812B
Anti-OR1N1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OR1N1 (olfactory receptor, family 1, subfamily N, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR