Anti-NADK2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13805CEA 350 EUR
ABGEAP13805C
Anti-NADK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NADK2 (NAD kinase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR