Anti-CSNK2A2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13804AEA 350 EUR
ABGEAP13804A
Anti-CSNK2A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CSNK2A2 (casein kinase 2, alpha prime polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR