Anti-S100A7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13794AEA 350 EUR
ABGEAP13794A
Anti-S100A7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: S100A7 (S100 calcium binding protein A7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR