Anti-DHRS7C Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13788CEA 350 EUR
ABGEAP13788C
Anti-DHRS7C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DHRS7C (dehydrogenase/reductase (SDR family) member 7C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR